Nicotinic acid (Niacin) ≥99.5% (by HPLC, titration analysis) matrix for MALDI-TOF/MS

Supplier: TCI America
Warning

N1103-1G N1103-5G
TCN1103-1GEA 38 USD
TCN1103-1G TCN1103-5G
Nicotinic acid (Niacin) ≥99.5% (by HPLC, titration analysis) matrix for MALDI-TOF/MS
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 14,06525

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.5 area%
Purity (Neutralization titration) min. 99.5 %
Melting point 236.0 to 240.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR