2-Nitroisophthalic acid ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

N1086-5G
TCN1086-5GEA 70 USD
TCN1086-5G
2-Nitroisophthalic acid ≥95.0% (by HPLC, titration analysis)
2-Nitroisophthalic acid
Formula: C₈H₅NO₆
MW: 211.1284 g/mol
MDL Number: MFCD00017007
CAS Number: 21161-11-5
EINECS: 244-253-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR