5-Norbornene-2-carboxamide (mixture of isomers) ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
CAS Number: 95-17-0
MDL Number: MFCD00187896
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method:
Other Product Names / Synonyms: Bicyclo[2.2.1]hept-5-ene-2-carboxamide
Formula:
C₈H₁₁NO Melting Pt: 188 °C |
MDL Number:
MFCD00187896 CAS Number: 95-17-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...