Naphthalene ≥98.0% (by GC)
Supplier: TCI America
N0885-25G
N0885-500G
TCN0885-25GEA
19.19
USD
TCN0885-25G
TCN0885-500G
Naphthalene ≥98.0% (by GC)
Naphthalene
CAS Number: 91-20-3
MDL Number: MFCD00001742
Molecular Formula: C10H8
Molecular Weight: 128.17
Purity/Analysis Method:
Formula:
C₁₀H₈ Boiling Pt: 218 °C Melting Pt: 81 °C Flash Pt: 79 °C |
MDL Number:
MFCD00001742 CAS Number: 91-20-3 UN: 1334 ADR: 4.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...