Naphthalene ≥98.0% (by GC)

Supplier: TCI America

N0885-25G N0885-500G
TCN0885-25GEA 19.19 USD
TCN0885-25G TCN0885-500G
Naphthalene ≥98.0% (by GC)
Naphthalene

CAS Number: 91-20-3 MDL Number: MFCD00001742 Molecular Formula: C10H8 Molecular Weight: 128.17 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 218 Melting point (°C): 81 Flash Point (°C): 79

Formula: C₁₀H₈
Boiling Pt: 218 °C
Melting Pt: 81 °C
Flash Pt: 79 °C
MDL Number: MFCD00001742
CAS Number: 91-20-3
UN: 1334
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4bd66d590350fdf511b0ab490eaee645121dc27&vid=e4bd66d590350fdf511b0ab490eaee645121dc27&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR