N-(1-Naphthyl)ethylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen) for biochemical research

Supplier: TCI America

Synonyms: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine

N0869-5G
TCN0869-5GEA 70.69 USD
TCN0869-5G
N-(1-Naphthyl)ethylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen) for biochemical research
N-2-Aminoethyl-1-naphthylamine dihydrochloride

[for Biochemical Research]
CAS Number: 1465-25-4
MDL Number: MFCD00012556
Molecular Formula: C12H14N2
Molecular Weight: 259.17
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 190


Product Categories: Bioscience, ROS/RNS Detection Reagents, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: Bratton-Marshall Reagent, Marshall's Reagent

Formula: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00012556
CAS Number: 1465-25-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28a49a68e65094ac0b2e419c05b010eebe04d8f4&vid=7f740e775a83e0795684293b8e8a09a40a81d02c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR