Naftopidil ≥98.0%
Supplier: TCI America
Synonyms:
1-[4-(2-Methoxyphenyl)piperazinyl]-3-(1-naphthyloxy)propan-2-ol
N0832-5G
TCN0832-5GEA
49.09
USD
TCN0832-5G
Naftopidil ≥98.0%
Naftopidil
CAS Number: 57149-07-2
MDL Number: MFCD00242741
Molecular Formula: C24H28N2O3
Molecular Weight: 392.50
Purity/Analysis Method:
Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 1-[4-(2-Methoxyphenyl)piperazinyl]-3-(1-naphthyloxy)propan-2-ol
Formula:
C₂₄H₂₈N₂O₃ MW: 392.50 g/mol Melting Pt: 127 °C |
MDL Number:
MFCD00242741 CAS Number: 57149-07-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...