Nigrosine, water soluble
Supplier: TCI America
Synonyms:
Acid Black 2
N0782-25G
TCN0782-25GEA
22.39
USD
TCN0782-25G
Nigrosine, water soluble
Nigrosine
CAS Number: 8005-03-6
Form: Crystal
Color: Black
Other Product Names / Synonyms: Acid Black 2
MDL Number:
MFCD00044681 CAS Number: 8005-03-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...