Ninhydrin in ethanol (contains Acetic Acid)

Supplier: TCI America
Warning

Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate

N0719-500ML
TCN0719-500MLEA 174.61 USD
TCN0719-500ML
Ninhydrin in ethanol (contains Acetic Acid)
Ninhydrin

[for TLC Stain]
CAS Number: 485-47-2
MDL Number: MFCD00003791
Molecular Formula: C9H6O4
Molecular Weight: 178.14
Form: Clear Liquid
Color: Slightly Pale Yellow

Formula: C₉H₆O₄
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 250…258 °C
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 13,06582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a6c8d9a6e311b58ece68338f20ac571e0a06b70&vid=33ed108b857a242ca191eafb985376f756fb7366&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR