Nalidixic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Danger

N0490-25G
TCN0490-025GEA 62.8 USD
TCN0490-025G
Nalidixic acid ≥95.0% (by titrimetric analysis)
Nalidixic acid

CAS Number: 389-08-2 MDL Number: MFCD00006884 Molecular Formula: C12H12N2O3 Molecular Weight: 232.24 Purity/Analysis Method: 95.0% (T) Form: Crystal Melting point (°C): 229


Product Categories: Bioscience, Synthetic Antimicrobial Ingredients Other Product Names / Synonyms: 1-Ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic Acid

Formula: C₁₂H₁₂N₂O₃
MW: 232.24 g/mol
Melting Pt: 229 °C
Storage Temperature: Ambient
MDL Number: MFCD00006884
CAS Number: 389-08-2
EINECS: 206-864-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99c4a1043ae287287916f90bbf026a0f68c03ac4&vid=99c4a1043ae287287916f90bbf026a0f68c03ac4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR