(R)-(+)-1-(1-Naphthyl)ethylamine ≥99.0%
Supplier: TCI America
Synonyms:
(R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine
CAS Number: 3886-70-2
MDL Number: MFCD00064114
Molecular Formula: C12H13N
Molecular Weight: 171.24
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (R)-(+)-alpha-(1-Aminoethyl)naphthalene
Formula:
C₁₂H₁₃N Boiling Pt: 80 °C Density: 1.07 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00064114 CAS Number: 3886-70-2 EINECS: 223-425-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...