(R)-(+)-1-(1-Naphthyl)ethylamine ≥99.0%

Supplier: TCI America

Synonyms: (R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine

N0482-5G N0482-1G
TCN0482-005GEA 122.04 USD
TCN0482-005G TCN0482-001G
(R)-(+)-1-(1-Naphthyl)ethylamine ≥99.0%
(R)-(+)-1-(1-Naphthyl)ethylamine

CAS Number: 3886-70-2 MDL Number: MFCD00064114 Molecular Formula: C12H13N Molecular Weight: 171.24 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 80 Flash Point (°C): 110 Specific Gravity (20/20): 1.07 Specific rotation [a]20/D: 80.5 deg (neat)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR) Other Product Names / Synonyms: (R)-(+)-alpha-(1-Aminoethyl)naphthalene

Formula: C₁₂H₁₃N
Boiling Pt: 80 °C
Density: 1.07 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00064114
CAS Number: 3886-70-2
EINECS: 223-425-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6356a7c71212cfde1ca9719f174931b63c0dfc38&vid=6356a7c71212cfde1ca9719f174931b63c0dfc38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR