Nerolidol (cis and trans mixture) ≥97.0%

Supplier: TCI America

Synonyms: 3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, 3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene, Nerolidol, 3,7,11-Trimethyldodeca-1,6,10-trien-3-ol

N0454-25ML
TCN0454-025MLEA 33.56 USD
TCN0454-025ML
Nerolidol (cis and trans mixture) ≥97.0%
Nerolidol (cis and trans mixture)

CAS Number: 7212-44-4 MDL Number: MFCD00008911 Molecular Formula: C15H26O Molecular Weight: 222.37 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 146 Flash Point (°C): 135 Specific Gravity (20/20): 0.88


Product Categories: Bioscience, Sesquiterpenes Other Product Names / Synonyms: 3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol

Formula: C₁₅H₂₆O
MW: 222.37 g/mol
Boiling Pt: 146 °C
Density: 0.88 (20 °C)
Flash Pt: 135 °C
MDL Number: MFCD00008911
CAS Number: 7212-44-4

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR