Norbornadiene rhodium(I) chloride dimer

Supplier: TCI America

Synonyms: Chloronorbornadiene rhodium(I) dimer, Bis(norbornadiene-mu-chlororhodium), Chloronorbornadiene Rhodium(I) Dimer

N0453-100MG
TCN0453-100MGEA 138.65 USD
TCN0453-100MG
Norbornadiene rhodium(I) chloride dimer
Norbornadiene rhodium(I) chloride dimer

CAS Number: 12257-42-0
MDL Number: MFCD00198060
Molecular Formula: C14H16Cl2Rh2
Molecular Weight: 460.99
Form: Crystal
Color: Very Pale Yellow


Other Product Names / Synonyms: Bis(norbornadiene-mu-chlororhodium), Chloronorbornadiene Rhodium(I) Dimer

Formula: C₁₄H₁₆Cl₂Rh₂
MDL Number: MFCD00198060
CAS Number: 12257-42-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b73f2fc7d16685070dbd64699db2cd62a5fa0fc&vid=d9053ff3c52394124b6d4eee7a12f04c3b4d30c7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR