p-Nitrophenyl phosphate DiTris salt ≥90.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Phosphoric acid 4-nitrophenyl ester Bis[tris(hydroxymethyl)aminomethane] salt, 4-Nitrophenyl phosphate Bis(tris) salt, p-Nitrophenyl phosphate DiTris Salt, 4-Nitrophenyl phosphatedi(tris) salt, pNPP 2Tris
[Substrate for Phosphatase]
CAS Number: 68189-42-4
MDL Number: MFCD00037009
Molecular Formula: C6H6NO6P
Molecular Weight: 461.36
Purity/Analysis Method:
Product Categories: Bioscience, Alkali Phosphatase Substrates
Other Product Names / Synonyms: Phosphoric Acid 4-Nitrophenyl Ester Bis[tris(hydroxymethyl)aminomethane] Salt, 4-Nitrophenyl Phosphate Bis(tris) Salt
Formula:
C₆H₆NO₆P MW: 461.36 g/mol |
MDL Number:
MFCD00037009 CAS Number: 68189-42-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...