5-Nonanol ≥97.0%

Supplier: TCI America

N0337-25ML
TCN0337-025MLEA 79 USD
TCN0337-025ML
5-Nonanol ≥97.0%
5-Nonanol

CAS Number: 623-93-8 MDL Number: MFCD00021944 Molecular Formula: C9H20O Molecular Weight: 144.26 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 193 Melting point (°C): 5 Flash Point (°C): 77 Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Dibutylcarbinol, 5-Nonyl Alcohol

Formula: C₉H₂₀O
Boiling Pt: 193 °C
Melting Pt: 5 °C
Density: 0.83 (20 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00021944
CAS Number: 623-93-8
EINECS: 210-820-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d518b690627a2142b933e45db8d19412a418b28&vid=2d518b690627a2142b933e45db8d19412a418b28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR