2-Nitrotoluene ≥99.0%

Supplier: TCI America
Danger

N0275-500G N0275-25G
TCN0275-500GEA 75 USD
TCN0275-500G TCN0275-25G
2-Nitrotoluene ≥99.0%
2-Nitrotoluene

CAS Number: 88-72-2 MDL Number: MFCD00007157 Molecular Formula: C7H7NO2 Molecular Weight: 137.14 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 222 Flash Point (°C): 106 Specific Gravity (20/20): 1.17

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 221.7
Melting Pt: -1
Density: 1.17 (20 °C)
Flash Pt: 106
Storage Temperature: Ambient
MDL Number: MFCD00007157
CAS Number: 88-72-2
EINECS: 201-853-3
UN: 1664
ADR: 6.1 II
Merck Index: 12,06748
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ab2b5c6a25724906d3f4fa126045a2f7aaabdbb&vid=c2f26448c920c2d0a4b2be4e2f39e72d39890f1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR