Nitromethane 55% ∼8.7 mol/L in methanol

Supplier: TCI America
Danger

N0239-25ML N0239-500ML
TCN0239-25MLEA 27 USD
TCN0239-25ML TCN0239-500ML
Nitromethane 55% ∼8.7 mol/L in methanol
Nitromethane

CAS Number: 75-52-5
MDL Number: MFCD00007400
Molecular Formula: CH3NO2
Molecular Weight: 61.04
Form: Clear Liquid
Color: Colorless

Formula: CH₃NO₂
Boiling Pt: 101.2 °C (1013 hPa)
Melting Pt: –29 °C
Density: 1.138 g/cm³ (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00007400
CAS Number: 75-52-5
EINECS: 200-876-6
UN: 1261
ADR: 3,II
Merck Index: 13,06644
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f828d6be0e34b3a0b357a9a031db4729dbe1fbb&vid=d30720f3f12ece19a1d9f6e3c9244ee49a815c97&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR