Benzyl nicotinate ≥98.0%

Supplier: TCI America
Warning

N0083-25G
TCN0083-025GEA 27 USD
TCN0083-025G TCN0083-100G
Benzyl nicotinate ≥98.0%
Benzyl nicotinate

CAS Number: 94-44-0 MDL Number: MFCD00023584 Molecular Formula: C13H11NO2 Molecular Weight: 213.24 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 189 Flash Point (°C): 113 Freezing point (°C): 22


Other Product Names / Synonyms: Nicotinic Acid Benzyl Ester

Formula: C₁₃H₁₁NO₂
Boiling Pt: 189 °C
Melting Pt: 24 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00023584
CAS Number: 94-44-0
EINECS: 202-332-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59fb701c7e5ff3fdd7b8e36abfc5ba20c92a5168&vid=59fb701c7e5ff3fdd7b8e36abfc5ba20c92a5168&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR