Nitromethane ≥98.0%

Supplier: TCI America
Danger

N0019-100ML
TCN0019-100MLEA 85.06 USD
TCN0019-100ML
Nitromethane ≥98.0%
Nitromethane

[for Spectrophotometry] CAS Number: 75-52-5 MDL Number: MFCD00007400 Molecular Formula: CH3NO2 Molecular Weight: 61.04 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 101 Melting point (°C): -29 Flash Point (°C): 35 Specific Gravity (20/20): 1.14 Lambda max.: 380 nm (H2O)

Formula: CH₃NO₂
MW: 61.04 g/mol
Boiling Pt: 101 °C
Melting Pt: –29 °C
Density: 1.14 (20 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00007400
CAS Number: 75-52-5
EINECS: 200-876-6
UN: 1261
ADR: 3,II
Merck Index: 13,06644
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=060d9e56e20cdf60744a25da8551367d47baf7f6&vid=83978bea00410fd590104763fd9c8c0b159bba18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR