Naphthalene ≥99.8% (by GC), zone refined: number of passes 23

Supplier: TCI America

N0004-1SAMPLE
TCN0004-ZONEEA 217 USD
TCN0004-ZONE
Naphthalene ≥99.8% (by GC), zone refined: number of passes 23
Naphthalene

CAS Number: 91-20-3
MDL Number: MFCD00001742
Molecular Formula: C10H8
Molecular Weight: 128.17
Purity/Analysis Method: >99.8% (GC)
Form: Crystal
Boiling point (°C): 218
Melting point (°C): 81
Flash Point (°C): 79

Formula: C₁₀H₈
Boiling Pt: 218 °C
Melting Pt: 81 °C
Flash Pt: 79 °C
MDL Number: MFCD00001742
CAS Number: 91-20-3
UN: 1334
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=293f8c036992f9a64faba920699a44914dee5abc&vid=1c3d39d7f39007085dc818810e5687b12b6ecd74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.8% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR