5-Methylbenzo[b]thiophene ≥98.0% (by GC)

Supplier: TCI America

M2974-1G M2974-5G
TCM2974-1GEA 116 USD
TCM2974-1G TCM2974-5G
5-Methylbenzo[b]thiophene ≥98.0% (by GC)
5-Methylbenzo[b]thiophene
Formula: C₉H₈S
MW: 148.23
Boiling Pt: 67°C
MDL Number: MFCD00052509
CAS Number: 14315-14-1
EINECS: 238-256-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 35.0 to 39.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR