Bisacrylamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS

M2877-25G M2877-500G
TCM2877-25GEA 29 USD
TCM2877-25G TCM2877-500G
Bisacrylamide ≥98.0% (by titrimetric analysis)
Bisacrylamide
Formula: (H₂C=CHCONH)₂CH₂
MW: 154.17 g/mol
Boiling Pt: 125
Melting Pt: 185
Flash Pt: 138
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Bromination Method) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR