JSH-23 ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Methyl-N1-(3-phenylpropyl)-1,2-phenylenediamine, JSH-23, 4-Methyl-N1-(3-phenylpropyl)benzene-1,2-diamine

M2734-100MG
TCM2734-100MGEA 437.3 USD
TCM2734-100MG TCM2734-25MG
JSH-23 ≥98.0% (by GC)
JSH-23
Formula: C₁₆H₂₀N₂
MW: 240.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09753595
CAS Number: 749886-87-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR