Methyl-4-bromo-3-nitrobenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Bromobenzoic acid methyl ester, Methyl 4-bromo-3-nitrobenzoate

M2596-1G M2596-5G
TCM2596-1GEA 20 USD
TCM2596-1G TCM2596-5G
Methyl-4-bromo-3-nitrobenzoate ≥98.0% (by GC)
Methyl 4-bromo-3-nitrobenzoate
Formula: C₈H₆BrNO₄
MDL Number: MFCD00553089
CAS Number: 2363-16-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR