2-Morpholino-8-phenylchromone ≥98.0%
Supplier: TCI America
Synonyms:
LY294002, LY 294002, 2-Morpholino-8-phenyl-4H-chromen-4-one, 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyran
M2410-25MG
TCM2410-25MGEA
229.1
USD
TCM2410-25MG
2-Morpholino-8-phenylchromone ≥98.0%
2-Morpholino-8-phenylchromone
CAS Number: 154447-36-6
MDL Number: MFCD00270881
Molecular Formula: C19H17NO3
Molecular Weight: 307.35
Purity/Analysis Method:
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Regenerative Medicine Research Reagents
Other Product Names / Synonyms: 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyran, LY 294002
Formula:
C₁₉H₁₇NO₃ Melting Pt: 184 °C |
MDL Number:
MFCD00270881 CAS Number: 154447-36-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...