Methylene blue solution in methanol for cell staining

Supplier: TCI America
Danger

Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

M2392-100ML
TCM2392-100MLEA 49 USD
TCM2392-100ML
Methylene blue solution in methanol for cell staining
Methylene blue
Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
MDL Number: MFCD00150008
CAS Number: 61-73-4
EINECS: 200-515-2
UN: 1230
ADR: 3,II
Merck Index: 14,06060

Order Now

Specification Test Results

Appearance Blue to Dark blue clear liquid to cloudy liquid
Absorbance min. 350 (664 nm)
Cell staining test Extent of staining same as reference

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR