Mycophenolic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

M2216-1G
TCM2216-1GEA 105.57 USD
TCM2216-1G
Mycophenolic acid ≥98.0% (by HPLC, titration analysis)
Mycophenolic acid

CAS Number: 24280-93-1
MDL Number: MFCD00036814
Molecular Formula: C17H20O6
Molecular Weight: 320.34
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 143
Storage Temperature: 0-10°C


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Inhibitors (for Pharmacological Research), Immunosuppressant Ingredients
Other Product Names / Synonyms: (E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic Acid

Formula: C₁₇H₂₀O₆
MW: 320.34 g/mol
Melting Pt: 143 °C
MDL Number: MFCD00036814
CAS Number: 24280-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff638733485239bbb98496f02d7abab64be020d2&vid=ff638733485239bbb98496f02d7abab64be020d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR