(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ∼18% ∼1.0 mol/L in dichloromethane for determination of the optical purity of alcohols and amines

Supplier: TCI America
Danger

M2215-5G
TCM2215-5GEA 573.3 USD
TCM2215-5G
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ∼18% ∼1.0 mol/L in dichloromethane for determination of the optical purity of alcohols and amines
(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Formula: C₁₀H₈ClF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00044400
CAS Number: 39637-99-5
UN: 3265
ADR: 8,III
Merck Index: 14,06280

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Optical purity (GC) min. 99.0 ee%
Concentration (Argentmetric titration) 18.0 to 20.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR