Myricetin ≥97.0%

Supplier: TCI America

Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one

M2131-500MG M2131-5G
TCM2131-500MGEA 121.4 USD
TCM2131-500MG TCM2131-5G
Myricetin ≥97.0%
Myricetin

CAS Number: 529-44-2 MDL Number: MFCD00006827 Molecular Formula: C15H10O8 Molecular Weight: 318.24 Purity/Analysis Method: 97.0% (HPLC) Form: Crystal Melting point (°C): 360


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Aromatic Polyketides, Polyphenols (Phytochemicals) Other Product Names / Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone

Formula: C₁₅H₁₀O₈
MW: 318.24 g/mol
Melting Pt: 360 °C
MDL Number: MFCD00006827
CAS Number: 529-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59c7d0df543240e1b6f58f00dbf31c257dce91ab&vid=59c7d0df543240e1b6f58f00dbf31c257dce91ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR