Myricetin ≥97.0%
Supplier: TCI America
Synonyms:
3,3',4',5,5',7-Hexahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one
M2131-500MG
M2131-5G
TCM2131-500MGEA
121.4
USD
TCM2131-500MG
TCM2131-5G
Myricetin ≥97.0%
Myricetin
CAS Number: 529-44-2
MDL Number: MFCD00006827
Molecular Formula: C15H10O8
Molecular Weight: 318.24
Purity/Analysis Method:
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone
Formula:
C₁₅H₁₀O₈ MW: 318.24 g/mol Melting Pt: 360 °C |
MDL Number:
MFCD00006827 CAS Number: 529-44-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...