2-Mercaptoethanol ≥99.0%

Supplier: TCI America

Synonyms: beta-Mercaptoethanol, 2-Hydroxyethyl mercaptan, β-Mercaptoethanol

M1948-25G M1948-5G
TCM1948-25GEA 60.7 USD
TCM1948-25G TCM1948-5G
2-Mercaptoethanol ≥99.0%
2-Mercaptoethanol

CAS Number: 60-24-2 MDL Number: MFCD00004890 Molecular Formula: C2H6OS Molecular Weight: 78.13 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 158 Melting point (°C): -100 Flash Point (°C): 74 Specific Gravity (20/20): 1.12


Product Categories: Bioscience, Disulfide Reducing Agents, Analytical Science, Reagents for Gel & Buffer Preparation, etc. (Reagents for Electrophoresis) Other Product Names / Synonyms: 2-Hydroxyethyl Mercaptan, Thioglycol

Formula: HSCH₂CH₂OH
MW: 78.14 g/mol
Boiling Pt: 157
Melting Pt: -100
Density: 1.12 (20 °C)
Flash Pt: 74 °C
MDL Number: MFCD00004890
CAS Number: 60-24-2
UN: 2966
ADR: 6.1 II
Merck Index: 13,05894
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bc34059936db0aeb8f2132e89a4c810b1b642d6&vid=8ac36b8922693d83965d30d9e71e4a80d37e34d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR