Methanol-[D₄] (99.8% D)
Supplier: TCI America
|
Danger
|
CAS Number: 811-98-3
MDL Number: MFCD00044637
Molecular Formula: CD4O
Molecular Weight: 36.07
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 66
Melting point (°C): -98
Flash Point (°C): 11
Specific Gravity (20/20): 0.89
Other Product Names / Synonyms: Tetradeuteromethanol, Methyl Alcohol-d4
Formula:
D₃COD Boiling Pt: 66 °C Melting Pt: –98 °C Density: 0.89 (20 °C) Flash Pt: 11 °C |
MDL Number:
MFCD00044637 CAS Number: 811-98-3 EINECS: 212-378-6 UN: 1230 ADR: 3,II |
Specification Test Results
Appearance | Colourless clear liquid |
Degree of deuteration | min. 99.8 atom%D |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...