3-Methylthiophene-2-carbonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1711-5G
TCM1711-5GEA 138.1 USD
TCM1711-5G
3-Methylthiophene-2-carbonyl chloride ≥98.0% (by GC, titration analysis)
3-Methylthiophene-2-carbonyl chloride

CAS Number: 61341-26-2 Molecular Formula: C6H5ClOS Molecular Weight: 160.62 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 220 Flash Point (°C): 90 Specific Gravity (20/20): 1.32


Other Product Names / Synonyms: 3-Methylthiophene-2-carbonyl Chloride

Formula: C₆H₅ClOS
Boiling Pt: 220 °C
Density: 1.32 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00130091
CAS Number: 61341-26-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24465020a25a170c8a8c057dcc682e58825e838c&vid=24465020a25a170c8a8c057dcc682e58825e838c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR