3-Methoxy-o-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 19500-02-8
MDL Number: MFCD06412568
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method:
Other Product Names / Synonyms: 1-Amino-3-methoxy-2-methylbenzene, 3-Methoxy-o-toluidine
Formula:
C₈H₁₁NO Density: 1.08 (20 °C) |
MDL Number:
MFCD06412568 CAS Number: 19500-02-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...