4-(4-Methylphenyl)-4-oxobutyric acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 4619-20-9
MDL Number: MFCD00020541
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method:
Other Product Names / Synonyms: 3-(4-Methylbenzoyl)propionic Acid, 4-(p-Tolyl)-4-oxobutyric Acid
Formula:
C₁₁H₁₂O₃ Melting Pt: 128 °C |
MDL Number:
MFCD00020541 CAS Number: 4619-20-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...