N-Methyl-D-aspartic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 6384-92-5
MDL Number: MFCD00004226
Molecular Formula: C5H9NO4
Molecular Weight: 147.13
Purity/Analysis Method:
Product Categories: Bioscience, Reagents for Other Disease Models, Biologically-modified Amino Acids
Other Product Names / Synonyms: N-Methyl-D-asparaginic Acid, Me-D-Asp-OH
Formula:
C₅H₉NO₄ Melting Pt: 178 °C |
MDL Number:
MFCD00004226 CAS Number: 6384-92-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...