2-Methyl-2-adamantanol ≥99.0%
Supplier: TCI America
Synonyms:
2-Hydroxy-2-methyladamantane, 2-Methyladamantan-2-ol
M1334-5G
M1334-25G
TCM1334-005GEA
109
USD
TCM1334-005G
TCM1334-025G
2-Methyl-2-adamantanol ≥99.0%
2-Methyl-2-adamantanol
CAS Number: 702-98-7
MDL Number: MFCD00074811
Molecular Formula: C11H18O
Molecular Weight: 166.26
Purity/Analysis Method:
Other Product Names / Synonyms: 2-Hydroxy-2-methyladamantane
Formula:
C₁₁H₁₈O Melting Pt: 217 °C |
MDL Number:
MFCD00074811 CAS Number: 702-98-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...