3-Methoxybenzonitrile ≥95.0% (by GC)

Supplier: TCI America

M1291-25G M1291-5G
TCM1291-25GEA 84.7 USD
TCM1291-25G TCM1291-5G
3-Methoxybenzonitrile ≥95.0% (by GC)
3-Methoxybenzonitrile

CAS Number: 1527-89-5 MDL Number: MFCD00001801 Molecular Formula: C8H7NO Molecular Weight: 133.15 Purity/Analysis Method: 95.0% (GC) Form: Crystal Boiling point (°C): 140 Flash Point (°C): 105 Freezing point (°C): 22


Other Product Names / Synonyms: 3-Cyanoanisole

Formula: C₈H₇NO
Boiling Pt: 140 °C
Flash Pt: 105 °C
MDL Number: MFCD00001801
CAS Number: 1527-89-5
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=324ef8f3f6c0ce6254e5a4976507d1265058c423&vid=a30288feb86edc11a7e8e6930b7ec89320b4b4b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR