1-Amino-2-methylbutane ≥98.0%

Supplier: TCI America

M1263-5ML M1263-25ML
TCM1263-005MLEA 98 USD
TCM1263-005ML TCM1263-25ML
1-Amino-2-methylbutane ≥98.0%
1-Amino-2-methylbutane

CAS Number: 96-15-1 MDL Number: MFCD00008147 Molecular Formula: C5H13N Molecular Weight: 87.17 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 97 Flash Point (°C): 3 Specific Gravity (20/20): 0.76


Other Product Names / Synonyms: 1-Amino-2-methylbutane

Formula: C₅H₁₃N
Boiling Pt: 97 °C
Density: 0.76 (20 °C)
Flash Pt: 3 °C
MDL Number: MFCD00008147
CAS Number: 96-15-1
EINECS: 202-483-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac12c8247d3a9ddd34d16f297b664970e566e04e&vid=ac12c8247d3a9ddd34d16f297b664970e566e04e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR