2-Mesitylenesulfonyl hydrazide ≥97.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 16182-15-3
MDL Number: MFCD00007585
Molecular Formula: C9H14N2O2S
Molecular Weight: 214.28
Purity/Analysis Method:
Other Product Names / Synonyms: 2,4,6-Trimethylbenzenesulfonyl Hydrazide
Formula:
C₉H₁₄N₂O₂S Melting Pt: 114 °C |
MDL Number:
MFCD00007585 CAS Number: 16182-15-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...