2-Methyl-2-pentanol ≥96.0% (by GC)

Supplier: TCI America

M1178-25ML M1178-5ML
TCM1178-25MLEA 140.2 USD
TCM1178-25ML TCM1178-5ML
2-Methyl-2-pentanol ≥96.0% (by GC)
2-Methyl-2-pentanol

CAS Number: 590-36-3 MDL Number: MFCD00004485 Molecular Formula: C6H14O Molecular Weight: 102.18 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Boiling point (°C): 122 Flash Point (°C): 30 Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: Dimethylpropylcarbinol

Formula: C₆H₁₄O
Boiling Pt: 122 °C
Density: 0.81 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00004485
CAS Number: 590-36-3
UN: 2560
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9790aae0d4a18a3d09a008585a59fdf90e219e9&vid=b9790aae0d4a18a3d09a008585a59fdf90e219e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR