4-Methoxyphenylphosphonic acid ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
4-Phosphonylanisole
M1155-1G
TCM1155-001GEA
311.8
USD
TCM1155-001G
4-Methoxyphenylphosphonic acid ≥95.0% (by titrimetric analysis)
4-Methoxyphenylphosphonic acid
CAS Number: 21778-19-8
MDL Number: MFCD00013948
Molecular Formula: C7H9O4P
Molecular Weight: 188.12
Purity/Analysis Method:
Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: 4-Phosphonylanisole
Formula:
C₇H₉O₄P MW: 188.12 g/mol |
MDL Number:
MFCD00013948 CAS Number: 21778-19-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...