Capsaicin 60.0% (by HPLC)

Supplier: TCI America

M1149-1G
TCM1149-001GEA 78.1 USD
TCM1149-001G
Capsaicin 60.0% (by HPLC)
Capsaicin

CAS Number: 404-86-4 MDL Number: MFCD00017259 Molecular Formula: C18H27NO3 Molecular Weight: 305.42 Purity/Analysis Method: 60.0% (HPLC) Form: Crystal Melting point (°C): 65 Flash Point (°C): 113


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Reagents for Pain & Itch Models, Reagents for Inflammation & Immunity Disorder Models, Alkaloids (Phytochemicals), Alkaloids (Others) Other Product Names / Synonyms: N-[(4-Hydroxy-3-methoxyphenyl)methyl]-8-methyl-6-nonenamide, 8-Methyl-N-vanillyl-6-nonenamide

Formula: C₁₈H₂₇NO₃
Boiling Pt: 210…220 °C (0.01 mmHg)
Melting Pt: 65 °C
Flash Pt: 113 °C
MDL Number: MFCD00017259
CAS Number: 404-86-4
EINECS: 206-969-8
UN: 2811
ADR: 6.1,III
Merck Index: 12,01811
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b99c18a2c4ef485b903ec4010aa61baceb9ac81&vid=4b99c18a2c4ef485b903ec4010aa61baceb9ac81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 60.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR