Methylcyclopentadiene dimer stabilized (so called)

Supplier: TCI America


Stabilization: Stabilised with Stabilised

M0920-25ML
TCM0920-025MLEA 20 USD
TCM0920-025ML
Methylcyclopentadiene dimer stabilized (so called)
Methylcyclopentadiene dimer

(stabilized with TBC) Form: Clear Liquid Color: Colorless Boiling point (°C): 200 Flash Point (°C): 53 Specific Gravity (20/20): 0.94 Molecular Formula: C12H16

Formula: C₁₂H₁₆
Boiling Pt: 200 °C
Density: 0.94 (20 °C)
Flash Pt: 53 °C
MDL Number: MFCD00082297
CAS Number: 26472-00-4
EINECS: 247-724-5
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=601be4e54f206e7ee8802309a01d27085885d02a&vid=129aafdf63318305f6a139a96022d46e78b2799c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR