2-Methylindoline ≥98.0%

Supplier: TCI America
Warning

M0871-25ML
TCM0871-025MLEA 54 USD
TCM0871-025ML
2-Methylindoline ≥98.0%
2-Methylindoline

CAS Number: 6872-06-6 MDL Number: MFCD00005706 Molecular Formula: C9H11N Molecular Weight: 133.19 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 230 Flash Point (°C): 93 Specific Gravity (20/20): 1.02

Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 228
Density: 1.02 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00005706
CAS Number: 6872-06-6
EINECS: 229-971-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ffc7156df5f5c5bfd6f89c65a70f104456eebcd&vid=9ffc7156df5f5c5bfd6f89c65a70f104456eebcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR