(±)-α-Methoxyphenylacetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 7021-09-2
MDL Number: MFCD00064217
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method:
Other Product Names / Synonyms: DL-O-Methylmandelic Acid
Formula:
C₉H₁₀O₃ Boiling Pt: 137 °C Melting Pt: 70 °C |
MDL Number:
MFCD00064217 CAS Number: 7021-09-2 EINECS: 230-300-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...