(S)-(+)-α-Methoxyphenylacetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
CAS Number: 26164-26-1
MDL Number: MFCD00064216
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method:
Formula:
C₉H₁₀O₃ Boiling Pt: 130 °C Melting Pt: 66 °C |
MDL Number:
MFCD00064216 CAS Number: 26164-26-1 EINECS: 247-492-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...