7-Amino-4-methylcoumarin ≥98.0%
Supplier: TCI America
Synonyms:
7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC
M0760-1G
M0760-5G
TCM0760-001GEA
136
USD
TCM0760-001G
TCM0760-5G
7-Amino-4-methylcoumarin ≥98.0%
7-Amino-4-methylcoumarin
[for HPLC Labeling]
CAS Number: 26093-31-2
MDL Number: MFCD00006868
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method:
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Boiling Pt: 352 Melting Pt: 226 Density: 1.377 g/cm³ (20 °C) |
MDL Number:
MFCD00006868 CAS Number: 26093-31-2 EINECS: 247-454-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...