MES sodium salt ≥97.0% for synthesis

Supplier: TCI America

Synonyms: Sodium 2-Morpholinoethanesulphonate, Sodium 2-morpholinoethane-1-sulfonate, Sodium 3-morpholin-1-ylethylsulphonate, Sodium 3-morpholin-1-ylethylsulfonate, 2-(N-Morpholino)ethanesulphonic acid sodium salt, Sodium 2-morpholinoethane-1-sulphonate, Sodium 2-Morpholinoethanesulfonate, 2-(N-Morpholino)ethanesulfonic acid sodium salt, MES-Na

M0737-25G
TCM0737-025GEA 98.67 USD
TCM0737-025G
MES sodium salt ≥97.0% for synthesis
MES sodium salt

CAS Number: 71119-23-8 MDL Number: MFCD00065473 Molecular Formula: C6H13NO4S Molecular Weight: 217.21 Purity/Analysis Method: 97.0% (N) Form: Crystal


Other Product Names / Synonyms: MES Sodium Salt, 2-Morpholinoethanesulfonic Acid Sodium Salt

Formula: C₆H₁₂NNaO₄S
MW: 217.22 g/mol
MDL Number: MFCD00065473
CAS Number: 71119-23-8
EINECS: 275-203-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6b8e7c7d6a27caa7b2b7c1849915dd913abaa8c&vid=e6b8e7c7d6a27caa7b2b7c1849915dd913abaa8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR